Learn More
Fine Science Tools Tungsten Needles, Tungsten Carbide, 1.2 cm, 10/pk
SDP
Catalog No. NC9463796
Encompass
Tungsten Needles Tungsten Needles - 0.5mm Rod Diameter/ Tip Diameter: 0.001mm/ Rod Diameter: 0.5mm/ Alloy: Tungsten Carbide/ Length: 1.2cm/ Unit: 10
Catalog No. NC9463796 Supplier Fine Science Tools Supplier No. 1013020
Tungsten Needles Tungsten Needles - 0.5mm Rod Diameter/ Tip Diameter: 0.001mm/ Rod Diameter: 0.5mm/ Alloy: Tungsten Carbide/ Length: 1.2cm/ Unit: 10