Learn More
Electron Microscopy Sciences Omniprobe Tungsten/Nickel Probe Tip
SDP
Catalog No. 5033759
Encompass
Custom tip designed with a nickel tube shank and tungsten tip. Tip radius is 0.5um with 13 degree taper angle for maximum lifetime service.
Catalog No. 50-337-59 Supplier Electron Microscopy Sciences Supplier No. 7596001
Custom tip designed with a nickel tube shank and tungsten tip. Tip radius is 0.5um with 13 degree taper angle for maximum lifetime service.