Learn More
LabScientific Tungsten Carbide Point Scriber/Etching Pen can mark and scribe steel, ceramic and glass. Glass slide etching pen. Fine point.
Catalog No. NC9587040
Encompass
The Tungsten Carbide Point Scriber/Etching Pen can mark and scribe steel, ceramic and glass. Point diameter: 1/16 in. (1.6mm)
Catalog No. NC9587040 Supplier LabScientific Supplier No. ETCHPEN
The Tungsten Carbide Point Scriber/Etching Pen can mark and scribe steel, ceramic and glass. Point diameter: 1/16 in. (1.6mm)