Learn More
Strem, An Ascensus Company CAS# 7440-33-7. 50g. Tungsten powder (99.9%). MFCD00011461
SDP
Catalog No. 5090116324
Encompass
CAS# 7440-33-7. 50g. Tungsten powder (99.9%). MFCD00011461. Molecular Weight: 183.85. Molecular Formula: W. Color/form: 12 micron Strem# 93-7456. http://www.strem.com/catalog/v/93-7456/
Catalog No. 50-901-16324 Supplier Strem, An Ascensus Company Supplier No. 93745650G
CAS# 7440-33-7. 50g. Tungsten powder (99.9%). MFCD00011461. Molecular Weight: 183.85. Molecular Formula: W. Color/form: 12 micron Strem# 93-7456. http://www.strem.com/catalog/v/93-7456/