Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten Silicide, W5Si3, MP Biomedicals

Catalog No. ICN224204 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN224204 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN224204 Supplier MP Biomedicals, Inc Supplier No. 0522420410

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12039-88-2
Molecular Formula Si2W
Molecular Weight (g/mol) 240.01
InChI Key WQJQOUPTWCFRMM-UHFFFAOYSA-N
Synonym tungsten disilicide, tungsten-silicide, tungsten silicide wsi2, tungsten silicide, w5si3
PubChem CID 16212546
IUPAC Name bis($l^{3}-silanylidyne)tungsten
SMILES [Si]#[W]#[Si]

Specifications

Quantity 10 g
Formula Weight 1003.51
Chemical Name or Material Tungsten Silicide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.