Learn More
Strem, An Ascensus Company CAS# 13283-01-7. 100g. Tungsten(VI) chloride (99.9%-W). MFCD00011463
SDP
Catalog No. 5090113029
Encompass
CAS# 13283-01-7. 100g. Tungsten(VI) chloride (99.9%-W). MFCD00011463. Molecular Weight: 396.57. Molecular Formula: WCl6. Color/form: purple xtl. Strem# 74-2350. http://www.strem.com/catalog/v/74-2350/
Catalog No. 50-901-13029 Supplier Strem, An Ascensus Company Supplier No. 742350100G
CAS# 13283-01-7. 100g. Tungsten(VI) chloride (99.9%-W). MFCD00011463. Molecular Weight: 396.57. Molecular Formula: WCl6. Color/form: purple xtl. Strem# 74-2350. http://www.strem.com/catalog/v/74-2350/