Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten(VI) ethoxide, 99.8% (metals basis), 5% w/v in ethanol

Catalog No. AA39170AE
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39170AE 100 mL
AA39170AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA39170AE Supplier Thermo Scientific Chemicals Supplier No. 039170AE
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 62571-53-3 | C12H30O6W | 454.21 g/mol

Tungsten(VI) ethoxide is used in nano-materials, thin films, display and imaging, optics and glasses. It is also used for x-ray screen phosphors, fireproofing fabrics and in gas sensors. It acts as an intermediate for sol-gel coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62571-53-3
Molecular Formula C12H30O6W
Molecular Weight (g/mol) 454.21
MDL Number MFCD00156526
InChI Key JHZFYDXXWXXVRM-UHFFFAOYSA-N
Synonym tungsten vi ethoxide, tungsten vi ethoxide w/v in ethanol, acmc-20ajad, wolfram vi hexaethoxide, ethanol, tungsten 6+ salt, tungsten 6+ hexakis ethoxide
PubChem CID 13828235
IUPAC Name ethanolate;tungsten
SMILES CCO[W](OCC)(OCC)(OCC)(OCC)OCC

Specifications

Color Brown to Yellow
Flash Point 17°C (63°F)
Odor Alcohol-like
Linear Formula W(OC2H5)6
Quantity 100 mL
UN Number UN1170
Sensitivity Moisture sensitive
Solubility Information Soluble in ethyl acetate,hydrocarbons and esters.
Formula Weight 454.21
Percent Purity 99.8%
Concentration or Composition (by Analyte or Components) 5% w/v in ethanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tungsten(VI) ethoxide
Show More Show Less
Hazard Category H225-H319-H335
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.