Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tungsten(VI) oxide

Catalog No. 5500865G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-008-65G 5 g
55-008-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55-008-65G Supplier Sigma Aldrich Supplier No. 5500865G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tungsten oxide (WO3) nanopowder is a metallic powder that can be prepared by the calcination of ammonium paratungstate (APT) in the temperature range of 560°C and 850°C. It can be produced by a variety of techniques, such as electrodeposition, sputtering, milling, and chemical processes.WO3 can be used for a variety of applications, such as electronics, optics, gas sensors and elecctrochromism.

Specifications

CAS 1314-35-8
Quantity 5 g
Molecular Formula O3W
Linear Formula WO3
MDL Number MFCD00011466
Synonym Tungstic anhydride
Molecular Weight (g/mol) 231.84
Density 7.16 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.