Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tungsten(VI) oxide

Catalog No. 95410100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
95-410-100G 100 g
95-410-500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 95-410-100G Supplier Sigma Aldrich Supplier No. 95410100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tungsten(VI) oxide (WO3) is a semiconducting material with excellent electrochromic properties. It is a cost efficient material with a high optical modulation. It can be synthesized by a variety of methods which include chemical vapor deposition, electrodeposition, and other sol-gel based coating processes.WO3 can be used for a wide range of applications such as electrochemical devices, photocatalysis, biological sensors, and other display devices.

Specifications

CAS 1314-35-8
Quantity 100 g
Molecular Formula O3W
Linear Formula WO3
MDL Number MFCD00011466
Synonym Tungstic anhydride
Molecular Weight (g/mol) 231.84
Percent Purity 99.9%
Density 7.16 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.