Learn More
Strem, An Ascensus Company CAS# 7440-33-7. 50m. Tungsten wire (99.95%). MFCD00011461
SDP
Catalog No. 5090116313
Encompass
CAS# 7440-33-7. 50m. Tungsten wire (99.95%). MFCD00011461. Molecular Weight: 183.85. Molecular Formula: W. Color/form: 0.127 mm dia. Strem# 93-7438. http://www.strem.com/catalog/v/93-7438/
Catalog No. 50-901-16313 Supplier Strem, An Ascensus Company Supplier No. 93743850MT
CAS# 7440-33-7. 50m. Tungsten wire (99.95%). MFCD00011461. Molecular Weight: 183.85. Molecular Formula: W. Color/form: 0.127 mm dia. Strem# 93-7438. http://www.strem.com/catalog/v/93-7438/