Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten wire basket; # turns, 9; ID(mm), 4; Ht(mm), 7; Wire leads(cm), 3.5; Wire dia(mm) 0.51, Thermo Scientific™

Catalog No. AA41177KD
Change view
Click to view available options
Quantity:
10 Pc.
50 Pc.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41177KD 50 Pc.
AA41177KK 10 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41177KD Supplier Thermo Scientific Chemicals Supplier No. 41177KD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Quantity 50 Pc.
Form Wire
Chemical Name or Material Tungsten wire basket, Turns: 9, ID: (4mm), Ht(7mm), Wire leads (3.5cm), Wire dia (0.51mm)
RTECSNumber YO7175000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.