Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tungsten wire, chromium plated, 1.8mm (0.07in) min dia, W 96.5+%, Cr 99.888% (metals basis)

Catalog No. AA41040DAT
Change view
Click to view available options
Quantity:
10 x 10 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41040DAT 10 x 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41040DAT Supplier Thermo Scientific™ Supplier No. 41040DAT
Add to Cart
Evaporation source, contacts and coatings
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 x 10 cm
Form Solid
Molecular Formula Tungsten wire thickness 1.3mm (0.05in)
MDL Number MFCD00011461
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.