Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tungstosilicic Acid Hydrate

Catalog No. T278650G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T278650G 50 g
T278610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. T278650G Supplier Sigma Aldrich Supplier No. T278650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tungstosilicic acid can be used as a reactant for:Preparation of silicotungstate butylmethylimidazolium bromide ionic liquid hybrid material.Synthesis of Sn(II)-substituted Keggin-type polyoxometalates as photocatalysts.Preparation of silicotungstic acid/organically modified silane proton-conducting membranes.Preparation of charge transfer compounds with Keggin anions.Preparation of catalyst for catalytic liquid-phase oxidation of xylene.

Specifications

CAS 12027-43-9
Molecular Formula H4O40SiW12 -+ xH2O
Linear Formula H4[Si(W3O10)4] -+ xH2O
Quantity 50 g
MDL Number MFCD00150392
Synonym Silicotungstic acid
Molecular Weight (g/mol) 2878.17 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.