Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tungstosilicic Acid Hydrate

Catalog No. 9539550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
95-395-50G 250 g
95-395-250G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 95-395-50G Supplier Sigma Aldrich Supplier No. 9539550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Preparation of silicotungstate butylmethylimidazolium bromide ionic liquid hybrid materialSynthesis of SnII-substituted Keggin-type polyoxometalates as photocatalystsPreparation of silicotungstic acid/organically modified silane proton-conducting membranesPreparation of charge transfer compounds with Keggin anionsPreparation of catalyst for catalytic liquid-phase oxidation of xylene

Specifications

CAS 12027-43-9
Molecular Formula H4O40SiW12 -+ xH2O
Linear Formula H4[Si(W3O10)4] -+ xH2O
MDL Number MFCD00150392
Quantity 250 g
Synonym Silicotungstic acid
Molecular Weight (g/mol) 2878.17 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.