Learn More
Enzo Life Sciences Tunicamycin, (10mg), CAS Number: 11089-65-9
SDP
Catalog No. NC0063615
Encompass_Preferred
GlcNAc phosphotransferase inhibitor. Nucleoside antibiotic. Inhibits protein N-glycosylation by blocking the transfer of N-acetylglucosamine 1-phosphate to dolichol monophosphate.  Induces ER-stress following the inhibition of N-linked glycosylation. Arrests cell cycle in late G1. Unstable at acidic pH. Formula: C38H62N4O16 (Tun B). MW: Avg. 840.0. CAS Number: 11089-65-9. MI: 14: 9819. Purity: ≥99% (TLC). Appearance: White solid to tan solid. Solubility: Soluble in DMSO (>10mg/ml) or methanol (5mg/ml; warm). Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: -20°C
Catalog No. NC0063615 Supplier Enzo Life Sciences Supplier No. BMLCC1040010
GlcNAc phosphotransferase inhibitor. Nucleoside antibiotic. Inhibits protein N-glycosylation by blocking the transfer of N-acetylglucosamine 1-phosphate to dolichol monophosphate.  Induces ER-stress following the inhibition of N-linked glycosylation. Arrests cell cycle in late G1. Unstable at acidic pH. Formula: C38H62N4O16 (Tun B). MW: Avg. 840.0. CAS Number: 11089-65-9. MI: 14: 9819. Purity: ≥99% (TLC). Appearance: White solid to tan solid. Solubility: Soluble in DMSO (>10mg/ml) or methanol (5mg/ml; warm). Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: -20°C