Learn More
Medchemexpress LLC HY-A0098 2mg Medchemexpress, Tunicamycin CAS:11089-65-9 Purity:>98%
SDP
Catalog No. 502029378
Encompass_Preferred
Medchemexpress, HY-A0098 2mg Tunicamycin CAS:11089-65-9 Tunicamycin is a mixture of homologous nucleoside antibiotic that inhibits N-linked glycosylation and blocks GlcNAc phosphotransferase (GPT). Tunicamycin causes accumulation of unfolded proteins in cell endoplasmic reticulum (ER) and induces ER stress, and causes blocking of DNA synthesis and cell cycle arrest in G1 phase. Tunicamycin inhibits gram-positive bacteria, yeasts, fungi, and viruses and has anti-cancer activity[2][3].Tunicamycin increases exosome release in cervical cancer cells[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9378 Supplier Medchemexpress LLC Supplier No. HYA00982MG
Medchemexpress, HY-A0098 2mg Tunicamycin CAS:11089-65-9 Tunicamycin is a mixture of homologous nucleoside antibiotic that inhibits N-linked glycosylation and blocks GlcNAc phosphotransferase (GPT). Tunicamycin causes accumulation of unfolded proteins in cell endoplasmic reticulum (ER) and induces ER stress, and causes blocking of DNA synthesis and cell cycle arrest in G1 phase. Tunicamycin inhibits gram-positive bacteria, yeasts, fungi, and viruses and has anti-cancer activity[2][3].Tunicamycin increases exosome release in cervical cancer cells[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.