Learn More
TARGETMOL CHEMICALS INC TUNICAMYCIN 5MG
SDP
Catalog No. 502581367
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tunicamycin is a mixture of homologous nucleoside antibiotic that inhibits N-linked glycosylation and blocks GlcNAc phosphotransferase (GPT). Tunicamycin inhibits gram-positive bacteria yeasts fungi and viruses and has anti-cancer activity. purity: 98%
Catalog No. 50-258-1367 Supplier TARGETMOL CHEMICALS INC Supplier No. T132295MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tunicamycin is a mixture of homologous nucleoside antibiotic that inhibits N-linked glycosylation and blocks GlcNAc phosphotransferase (GPT). Tunicamycin inhibits gram-positive bacteria yeasts fungi and viruses and has anti-cancer activity. purity: 98%