Learn More
Abcam Tunicamycin, Protein glycosylation inhibitor, 10MG
Catalog No. NC0634537
Encompass_Preferred
MW 816.9 Da, Purity >98%. Nucleoside antibiotic that inhibits protein glycosylation. Inhibits GlcNAc phosphotransferase (GPT) and inhibits the transfer of N-acetylglucosamine-1-phosphate from UDP-N-acetylglucosamine to dolichol phosphate in the first step of glycoprotein synthesis. Inhibits N-linked glycosylation and blocks the formation of N-glycosidic protein-carbohydrate linkages. Active in vitro against Gram-positive bacteria, yeasts, fungi and viruses.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0634537 Supplier Abcam Supplier No. AB12029610MG
MW 816.9 Da, Purity >98%. Nucleoside antibiotic that inhibits protein glycosylation. Inhibits GlcNAc phosphotransferase (GPT) and inhibits the transfer of N-acetylglucosamine-1-phosphate from UDP-N-acetylglucosamine to dolichol phosphate in the first step of glycoprotein synthesis. Inhibits N-linked glycosylation and blocks the formation of N-glycosidic protein-carbohydrate linkages. Active in vitro against Gram-positive bacteria, yeasts, fungi and viruses.
The product is subject to the following: Abcam Restricted Use Statement