Learn More
Cayman Chemical Tunicamycin Mixture, Purity: ≥95% (mixture of congeners), CAS: 11089-65-9, 10 mg
SDP
Catalog No. 502951483
Encompass_Preferred
A mixture of tunicamycins with variable N-acyl fatty acid chain lengths; inhibitor of HexNAc phosphotransferases and peptidoglycan biosynthesis in bacteria; inhibitor of GPT and glycosylation in eukaryotes; sensitizes EGFR inhibitor-resistant U251 and Bx/PC3 cells to irradiation
Catalog No. 50-295-1483 Supplier Cayman Chemical Supplier No. 1144510 MG
A mixture of tunicamycins with variable N-acyl fatty acid chain lengths; inhibitor of HexNAc phosphotransferases and peptidoglycan biosynthesis in bacteria; inhibitor of GPT and glycosylation in eukaryotes; sensitizes EGFR inhibitor-resistant U251 and Bx/PC3 cells to irradiation