Learn More
Medchemexpress LLC Tunicamycin V | 66054-36-2 | MFCD00040700 | 1 MG
SDP
Catalog No. 5000417236
Encompass_Preferred
Tunicamycin V (Tunicamycin A) is a nucleoside natural product that inhibits bacterial phospho-N-acetylmuramyl-pentapeptide transferase (MraY) with an IC50 of 0.35 μM. It exhibits antibacterial activities. Tunicamycins are natural products isolated from the fermentation broths of *Streptomyces lysosuperficus*, possessing various biological properties, including antibacterial, antiviral, antifungal, and antitumor activities. They strongly inhibit UDP-N-acetylglucosamine (GlcNAc): polyprenol phosphate translocase, an enzyme responsible for the first N-acetylglucosamination of the N-linked glycopeptide in the endoplasmic reticulum (ER).
  • Inhibits bacterial phospho-N-acetylmuramyl-pentapeptide transferase (MraY)
  • Possesses antibacterial, antiviral, antifungal, and antitumor activities
  • Strongly inhibits UDP-N-acetylglucosamine (GlcNAc): polyprenol phosphate translocase
  • Nucleoside natural product
Catalog No. 50-004-17236 Supplier Medchemexpress LLC Supplier No. HYN83951MG
Tunicamycin V (Tunicamycin A) is a nucleoside natural product that inhibits bacterial phospho-N-acetylmuramyl-pentapeptide transferase (MraY) with an IC50 of 0.35 μM. It exhibits antibacterial activities. Tunicamycins are natural products isolated from the fermentation broths of *Streptomyces lysosuperficus*, possessing various biological properties, including antibacterial, antiviral, antifungal, and antitumor activities. They strongly inhibit UDP-N-acetylglucosamine (GlcNAc): polyprenol phosphate translocase, an enzyme responsible for the first N-acetylglucosamination of the N-linked glycopeptide in the endoplasmic reticulum (ER).
  • Inhibits bacterial phospho-N-acetylmuramyl-pentapeptide transferase (MraY)
  • Possesses antibacterial, antiviral, antifungal, and antitumor activities
  • Strongly inhibits UDP-N-acetylglucosamine (GlcNAc): polyprenol phosphate translocase
  • Nucleoside natural product