Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Tuning Fork and Striker Set
GREENER_CHOICESCI_ED

Catalog No. S160133 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
3 to 9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S160133 3 to 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S160133 Supplier Supertek™ Supplier No. PH36121/10SET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Supertek™ Tuning Fork and Striker Set features a top-quality aluminum tuning fork with a frequency of 512 Hz and a 7 in. striker for optimal performance.

  • Striker has a wooden handle and rubber ball at the top.
  • Packaged together in a plastic bag.

Specifications

Grade 3 to 9
Format Kit
Includes 1 Aluminum tuning fork of 512 Hz and 1 7 in. striker
Languages English
Subject Physics - Sound and Light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.