Learn More
Crescent Chemical Co Inc TUNING/MASSCALIB 3 11COMP100ML
SDP
Catalog No. 5068578
Encompass_Preferred
TuningMass Calibration Multi-Element Mix 3, 11 components @10ugmL in 5% HNO3 CAS Size - 500mL Catalog Number - CC5654
Catalog No. 50-685-78 Supplier Crescent Chemical Co Inc Supplier No. CTCC5654
TuningMass Calibration Multi-Element Mix 3, 11 components @10ugmL in 5% HNO3 CAS Size - 500mL Catalog Number - CC5654