Learn More
Perkin Elmer US LLC Tuning Solution 1, 125 mL
Catalog No. NC1626671
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 2% HNO3/5% HCl 10 µg/mL: Ba, Be, Ce, Co, ln, Li, Mg, Pb, Rh, Tl, U, Y
Catalog No. NC1626671 Supplier Perkin Elmer US LLC Supplier No. N9303843
Concentration of Element includes: Matrix Concentration 2% HNO3/5% HCl 10 µg/mL: Ba, Be, Ce, Co, ln, Li, Mg, Pb, Rh, Tl, U, Y