Learn More
AccuStandard, Inc TUNING STD 5X1 ML
SDP
Catalog No. NC9973905
Encompass
GC/MS Tuning Standard for EPA Method 624/625. 100 mL. 10 ?g/mL in 2-5% Nitric Acid.
Catalog No. NC9973905 Supplier AccuStandard, Inc Supplier No. M625TSPAK
GC/MS Tuning Standard for EPA Method 624/625. 100 mL. 10 ?g/mL in 2-5% Nitric Acid.