Learn More
HANNA INSTRUMENTS INC Turbidity Calibration Standards
SDP
Supplier:  HANNA INSTRUMENTS INC HI9870311
Encompass
The HI98703-11 Turbidity Calibration Standards provide a simple solution to calibrating and validating turbidity meters that have a 0 to 1000 NTU range. Hanna turbidity calibration standards are prepared from NIST traceable primary standard reference materials and come supplied with a certificate of analysis. The certificate of analysis provides the lot number, reference values, and expiration date for traceability when certifying the appropriate turbidity meter.Supplied with certificate of analysisAMCO AEPA-1 primary turbidity standardsReusable with long shelf life stability
Catalog No. NC1760793