Learn More
Associates of Cape Cod Inc TURBIDIMETRIC LAL, 5 ML 25PK
SDP
Catalog No. NC0774125
Encompass
PYROTELL-T TURBIDIMETRIC LAL, SENSITIVE TO 0.001 EU/mL WHEN USED WITH ACC TUBE READERS, 5 mL/VIAL, 25 VIALS/PACK
Catalog No. NC0774125 Supplier Associates of Cape Cod Inc Supplier No. T005125
PYROTELL-T TURBIDIMETRIC LAL, SENSITIVE TO 0.001 EU/mL WHEN USED WITH ACC TUBE READERS, 5 mL/VIAL, 25 VIALS/PACK