Learn More
Turbidity 0.5 NTU Calibration Standard, Formazin Certified Reference Material, MilliporeSigma™ Supelco™
Catalog No. 111001787 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-1787 100 mL
11-100-1776 500 mL
11-100-1769 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-100-1787 Supplier MilliporeSigma Supelco Supplier No. TURB05100ML
Add to Cart
Add to Cart
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL
Grade Certified Reference Material