Learn More
ERA WATERS CORP TURBIDITY PT
SDP
Catalog No. NC0668604
Encompass
One 15 mL screw-cap vial yields up to 1 liter after dilution. Use with nephelometric methods.
Catalog No. NC0668604 Supplier ERA WATERS CORP Supplier No. 893
One 15 mL screw-cap vial yields up to 1 liter after dilution. Use with nephelometric methods.