Learn More
Aqua Solutions StablCal Turbidity Standard, 200 NTU, Bottle, 1 L
SDP
Catalog No. NC2212634
Encompass
The Hach StablCal Turbidity Standard 200 NTU is a 1 liter bottle of stabilized formazin turbidity standard with a nominal value of 200 NTU. It is used for calibration and verification of turbidimeters measuring low to moderate turbidity ranges. This ready-to-use standard has a two year shelf life and requires no dilution. It is formulated to minimize settling and provides a stable reference value traceable to AMCO standards. The 200 NTU standard is ideal for drinking water, wastewater, and industrial water applications.
This item is not returnable. View return policy
Catalog No. NC2212634 Supplier Aqua Solutions Supplier No. HA2660149
The Hach StablCal Turbidity Standard 200 NTU is a 1 liter bottle of stabilized formazin turbidity standard with a nominal value of 200 NTU. It is used for calibration and verification of turbidimeters measuring low to moderate turbidity ranges. This ready-to-use standard has a two year shelf life and requires no dilution. It is formulated to minimize settling and provides a stable reference value traceable to AMCO standards. The 200 NTU standard is ideal for drinking water, wastewater, and industrial water applications.