Learn More
Medchemexpress LLC Turbinaric acid | 56882-00-9 | 98.1% | 400.64 | 25 MG
SDP
Catalog No. 5000439487
Encompass_Preferred
Turbinaric acid is a type of secosqualene carboxylic acid that exhibits cytotoxicity against both mouse melanoma cells and human colon cancer cells. It serves as a valuable chemical marker for *T. conoides* and is utilized in research concerning melanoma and colon cancer.
  • Secosqualene carboxylic acid
  • Cytotoxicity against murine melanoma cells
  • Cytotoxicity against human colon carcinoma cells
  • Useful as a chemical marker for *T. conoides*
  • Supports research on melanoma and colon cancer
Catalog No. 50-004-39487 Supplier Medchemexpress LLC Supplier No. 25MGHY111604
Add to Cart
Add to Cart
Turbinaric acid is a type of secosqualene carboxylic acid that exhibits cytotoxicity against both mouse melanoma cells and human colon cancer cells. It serves as a valuable chemical marker for *T. conoides* and is utilized in research concerning melanoma and colon cancer.
  • Secosqualene carboxylic acid
  • Cytotoxicity against murine melanoma cells
  • Cytotoxicity against human colon carcinoma cells
  • Useful as a chemical marker for *T. conoides*
  • Supports research on melanoma and colon cancer