Learn More
MSC Kranzle Repair Kit, Industrial Dirt Killer Rotary Nozzle, Size 3.5
Catalog No. 502835911
Encompass
This repair kit is designed for Kränzle Industrial Dirt Killer Rotary Nozzles, allowing for easy and complete internal component replacement. It includes all necessary parts, such as the rotor, seat, and O-ring, to restore the nozzle to like-new performance. Installation is straightforward and can typically be completed in just five minutes, ensuring efficient maintenance of your equipment.
  • Replaces all internal parts
  • Includes rotor, seat, and O-ring
  • Restores nozzle to like-new condition
  • Simple installation
  • Quick repair, typically completed in five minutes
  • Ensures correct fit for specific nozzle models
Catalog No. 50-283-5911 Supplier MSC Supplier No. 57897944
This repair kit is designed for Kränzle Industrial Dirt Killer Rotary Nozzles, allowing for easy and complete internal component replacement. It includes all necessary parts, such as the rotor, seat, and O-ring, to restore the nozzle to like-new performance. Installation is straightforward and can typically be completed in just five minutes, ensuring efficient maintenance of your equipment.
  • Replaces all internal parts
  • Includes rotor, seat, and O-ring
  • Restores nozzle to like-new condition
  • Simple installation
  • Quick repair, typically completed in five minutes
  • Ensures correct fit for specific nozzle models