Learn More
BPS Bioscience Inc Turbo Nuclease, 250 units/μl, recombinant, ≥99% purity, 27 kDa, formulated with 50 mM Tris-HCl, pH 8.0, 50 mM NaCl, 5 mM MgCl₂, 50% glycerol
SDP
Catalog No. NC0598876
Encompass
Turbo Nuclease, 250 units/μl, recombinant, ≥99% purity, 27 kDa, formulated with 50 mM Tris-HCl, pH 8.0, 50 mM NaCl, 5 mM MgCl₂, 50% glycerol
Catalog No. NC0598876 Supplier BPS Bioscience Inc Supplier No. 50310
Turbo Nuclease, 250 units/μl, recombinant, ≥99% purity, 27 kDa, formulated with 50 mM Tris-HCl, pH 8.0, 50 mM NaCl, 5 mM MgCl₂, 50% glycerol