Learn More
ETON BIOSCIENCE INC TEV Protease (TurboTEV), 1 mg (10,000 Units)
Catalog No. NC9944529
Encompass
TurboTEV Protease contains an enhanced form of a catalytic fragment of the N1a protein of Tobacco etch virus (TEV), a cysteine protease that recognizes the cleavage site of Glu-Asn-Leu-Tyr-Phe-Gln-Gly and cleaves between Gln and Gly. TurboTEV Protease is a restriction grade protease that has a robust activity at 4oC with high specificity and great stability.
Catalog No. NC9944529 Supplier ETON BIOSCIENCE INC Supplier No. 1500020012
TurboTEV Protease contains an enhanced form of a catalytic fragment of the N1a protein of Tobacco etch virus (TEV), a cysteine protease that recognizes the cleavage site of Glu-Asn-Leu-Tyr-Phe-Gln-Gly and cleaves between Gln and Gly. TurboTEV Protease is a restriction grade protease that has a robust activity at 4oC with high specificity and great stability.