Learn More
OriGene TurboFectin 8.0 Transfection Reagent (5 mL in 5 vials)
SDP
Catalog No. 5000600185
Encompass
TurboFectin Transfection Reagent (5 mL in 5 vials)Components: Five Vials TurboFectin Transfection Reagents (1 mL per vial) Application GuideFeatures: Wide cell spectrum: valided on >100 cell lines and primary cells, view list, Very low cell toxicity, Simplified use: compatible with serum and antibiotics in the mediaQuality Control: Each batch of TurboFectin Transfection Reagent has been tested and yields at least 8 ng of luciferase per 35 mm well after transfection into NIH3T3 cells. TurboFectin reagent is supplied in ethanol to ensure sterilityStability: The transfection reagent is stable for one (1) year from date of shipping when stored at +4°C.Storage: Upon receipt, please store the TurboFectin Transfection Reagent at +4°C.Type: Transfection Reagent
Catalog No. 50-006-00185 Supplier OriGene Supplier No. TF81005
TurboFectin Transfection Reagent (5 mL in 5 vials)Components: Five Vials TurboFectin Transfection Reagents (1 mL per vial) Application GuideFeatures: Wide cell spectrum: valided on >100 cell lines and primary cells, view list, Very low cell toxicity, Simplified use: compatible with serum and antibiotics in the mediaQuality Control: Each batch of TurboFectin Transfection Reagent has been tested and yields at least 8 ng of luciferase per 35 mm well after transfection into NIH3T3 cells. TurboFectin reagent is supplied in ethanol to ensure sterilityStability: The transfection reagent is stable for one (1) year from date of shipping when stored at +4°C.Storage: Upon receipt, please store the TurboFectin Transfection Reagent at +4°C.Type: Transfection Reagent