Learn More
Proteintech Group Inc TurboGFP-Trap Magnetic Agarose
SDP
Supplier:  Proteintech Group Inc TBTMA20
Encompass
TurboGFP-Trap Magnetic Agarose is an affinity reagent for immunoprecipitation (IP) of TurboGFP fusion proteins.
Catalog No. 50-225-8885