Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Miller™ TurboLite™ Flash Personal Fall Limiters

Catalog No. 19085725
Change view
Click to view available options
Length (English):
6 ft.
9 ft.
Connector Types:
Locking Snap Hook and Aluminum Captive Eye Carabiner
Quick-Connect Harness Connector and Aluminum Rebar Hook
Quick-Connect Harness Connector and Steel Snap Hook
Quick-Connect Harness Connector, Aluminum Captive Eye Carabiner, Steel Tie-Back Snap Hook (5 K)
Steel Locking Snap Hook, Steel Twist-Lock Carabiner
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Connector Types
19-085-725 9 ft. Quick-Connect Harness Connector, Aluminum Captive Eye Carabiner, Steel Tie-Back Snap Hook (5 K)
19-085-722 6 ft. Quick-Connect Harness Connector and Aluminum Rebar Hook
19-085-723 6 ft. Quick-Connect Harness Connector and Steel Snap Hook
19-085-724 6 ft. Locking Snap Hook and Aluminum Captive Eye Carabiner
19-085-721 6 ft. Steel Locking Snap Hook, Steel Twist-Lock Carabiner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-085-725 Supplier Honeywell Miller™ Supplier No. MFLAFT1/9FT

missing translation for 'validMainframeMsg'

Designed specifically for those who work at height and are exposed to arc flash hazards, welding, or who are working near high heat areas.

  • Engineered to deliver the assurance that is expected from Miller™
  • TurboLite™ is an all-in-one solution for Arc flash and hot work applications
  • Provides versatility, reliability, and maximum safety
  • High impact housing with integral shock absorber
  • Rated for up to a 420 lb. (189 kg) weight capacity for all connections down to foot level
  • Meets the most stringent safety standards, including ANSI Z359.14 and ASTM F887
  • Available in a wide range of lengths and configurations including single or twin versions, tie-back options, and products specifically designed for users working in bucket trucks
TRUSTED_SUSTAINABILITY

Specifications

Color Black, Yellow
Product Type Personal Fall Limiter
Webbing Material Dupont™ Kevlar™™/Nomex™
For Use With (Application) Arc Flash and Hot Work Applications
Quantity 1 Ea.
Width (English) 1 in.
Leg Configuration Single
Capacity (English) 420 lb.
Certifications/Compliance ANSI A10.32, ANSI Z359.14, ASTM F877, OSHA 1910.140, and OSHA 1926.502
Connector Types Quick-Connect Harness Connector, Aluminum Captive Eye Carabiner, Steel Tie-Back Snap Hook (5 K)
Length (English) 9 ft.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.