Learn More
Perkin Elmer US LLC TurboMatrix HS PC Cable, Software Communication
Catalog No. 502103990
Encompass_Preferred
TurboMatrix HS PC Control Software provides full external control of a TurboMatrix headspace sampler from a personal computer running under Windows 95, 98 or NT. Methods and sequences can be generated, stored, edited, printed and executed.
Catalog No. 50-210-3990 Supplier Perkin Elmer US LLC Supplier No. M0417035
TurboMatrix HS PC Control Software provides full external control of a TurboMatrix headspace sampler from a personal computer running under Windows 95, 98 or NT. Methods and sequences can be generated, stored, edited, printed and executed.