Learn More
Perkin Elmer US LLC TurboMatrix Thermal Desorber Air Dryer
Catalog No. 502103951
Encompass_Preferred
Single-tube, manual-pneumatics model, upgradeable to the automated version. Includes a separate trap-clean-and-test function, which saves time and protects the GC column and detector.
Catalog No. 50-210-3951 Supplier Perkin Elmer US LLC Supplier No. N6710048
Single-tube, manual-pneumatics model, upgradeable to the automated version. Includes a separate trap-clean-and-test function, which saves time and protects the GC column and detector.