Learn More
OHAUS TurboMix Attachment
Catalog No. 01804353 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-353 Vortex Attachment
1 options
Catalog No. 01-804-353 Supplier OHAUS Supplier No. 83041390
Add to Cart
Add to Cart
Permanently mounts to most existing Vortex Genie 2 mixers and Vortex Genie Pulse.
  • It provides extremely vigorous mixing of up to twelve 1.5 mL or 2.0 mL microtubes (snap-top or screw-cap) through a combination of vortexing and impact collision
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Vortex-Genie 2 OR Vortex-Genie Pulse