Learn More
Research Products International Corp Turbo Mix Attachment for Vortex-Genie, Holds 12 Tubes (1.5 ml - 2.0 ml)
SDP
Catalog No. 50212390
Encompass
The New and Improved patented TurboMix attachment permanently mounts to most existing Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse to provide extremely vigorous mixing of up to twelve 1.5ml or 2.0ml microtubes (snap-top or screw-cap) through a combination of vortexing and impact collision. This system is ideal for aggressive and rapid dissolution, resuspension or mixing of samples and is also convenient and effective for cell disruption in glass bead procedures.
Catalog No. 50-212-390 Supplier Research Products International Corp Supplier No. 155304
The New and Improved patented TurboMix attachment permanently mounts to most existing Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse to provide extremely vigorous mixing of up to twelve 1.5ml or 2.0ml microtubes (snap-top or screw-cap) through a combination of vortexing and impact collision. This system is ideal for aggressive and rapid dissolution, resuspension or mixing of samples and is also convenient and effective for cell disruption in glass bead procedures.