Learn More
Accelagen TURBONUCLEASE 50 000 UNITS
Catalog No. NC0298896
Encompass
TurboNuclease is a recombinant Serratia macescens endonuclease (coded by the same gene as Bezonase). TurboNuclease degrades both DNA and RNA to oligonucleotides. This product is in low endotoxin, free of protease activity, and >99% pure with exceptionally high specific activity.
Catalog No. NC0298896 Supplier Accelagen Supplier No. N0103M
TurboNuclease is a recombinant Serratia macescens endonuclease (coded by the same gene as Bezonase). TurboNuclease degrades both DNA and RNA to oligonucleotides. This product is in low endotoxin, free of protease activity, and >99% pure with exceptionally high specific activity.