Learn More
Nacalai USA Inc TURBOTEV (TEV PROTEASE) 2MG/ML
Catalog No. NC9973044
Encompass
TurboTEV doesn't require any special buffer and can be used in a buffer most suitable for the target protein, It is a 52kDa protein with both GST and His tags so it can be removed by either Ni-chelating or Glutathione resin.
Catalog No. NC9973044 Supplier Nacalai USA Inc Supplier No. NU0102S
TurboTEV doesn't require any special buffer and can be used in a buffer most suitable for the target protein, It is a 52kDa protein with both GST and His tags so it can be removed by either Ni-chelating or Glutathione resin.