Learn More
Medchemexpress LLC HY-N2548 5mg Medchemexpress, Turkesterone CAS:41451-87-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN25485MG
Encompass_Preferred
Medchemexpress, HY-N2548 5mg Turkesterone CAS:41451-87-0 Turkesterone, isolated from Ajuga turkestanica, is a potent ecdysteroid. Turkesterone acts as an ecdysteroid receptor (EcR) agonist in some insect systems. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1517