Learn More
Innovative Research Inc Turkey Red Blood Cells Packed 5% 100ml
Catalog No. 502034858
Encompass_Preferred
Turkey Red Blood Cells from Innovative Research are manufactured using whole blood collections from Turkeys located in the USA. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4858 Supplier Innovative Research Inc Supplier No. ITKRBC5P100ML
Turkey Red Blood Cells from Innovative Research are manufactured using whole blood collections from Turkeys located in the USA. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.