Learn More
Innovative Research Inc Turkey Red Blood Cells Packed 5% 50ml
Catalog No. 502034864
Encompass_Preferred
Turkey Red Blood Cells from Innovative Research are manufactured using whole blood collections from turkeys. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 5% suspension in phosphate buffered saline (PBS). Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4864 Supplier Innovative Research Inc Supplier No. ITKRBC5P50ML
Turkey Red Blood Cells from Innovative Research are manufactured using whole blood collections from turkeys. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 5% suspension in phosphate buffered saline (PBS). Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.