Learn More
CTL Scientific Supply Corp TURMERIC PAPER,200 STRIPS
SDP
Catalog No. 501173306 Shop All CTL Scientific Supply Products
Encompass_Preferred
TURMERIC PAPER - box of 200 strips (each strip 20 x 70 mm). The test paper allows the quick and easy detection of boric acid and borates. Limit of sensitivity: 20 mg/l boron
Catalog No. 50-117-3306 Supplier CTL Scientific Supply Corp Supplier No. 90747
TURMERIC PAPER - box of 200 strips (each strip 20 x 70 mm). The test paper allows the quick and easy detection of boric acid and borates. Limit of sensitivity: 20 mg/l boron