Learn More
BURKLE INC Pressure sprayer Turn 'n' Spray, 50.72 oz. (1500 ml)
Catalog No. 5000025747
Encompass_Preferred
The Turn'n'Spray overhead pressure sprayer allows continuous spraying, even overhead or at an angle, thanks to its convenient 360° spray function.
Conventional pressure sprayers cannot spray continuously at an angle or overhead because they stop spraying as soon as the suction tube is out of the liquid. Not so with our Turn'n'Spray overhead pressure sprayer. It allows you to spray practically and comfortably from all directions, reaching hard-to-reach areas easily and without contortions. The pressure sprayer allows you to work comfortably, time-saving and uninterrupted.
Catalog No. 50-000-25747 Supplier BURKLE INC Supplier No. 03090120
The Turn'n'Spray overhead pressure sprayer allows continuous spraying, even overhead or at an angle, thanks to its convenient 360° spray function.
Conventional pressure sprayers cannot spray continuously at an angle or overhead because they stop spraying as soon as the suction tube is out of the liquid. Not so with our Turn'n'Spray overhead pressure sprayer. It allows you to spray practically and comfortably from all directions, reaching hard-to-reach areas easily and without contortions. The pressure sprayer allows you to work comfortably, time-saving and uninterrupted.