Learn More
Grainger Turnbuckle Body 1 600 lb Load Cap 4 in L
Catalog No. 502767227
Encompass_Preferred
The Loos & Co. Turnbuckle Body is a forged steel turnbuckle with a body diameter of 1-1/16 inches. It has a working load limit of 3,200 pounds and meets ASTM A668 specifications. The turnbuckle has a drop forged body with swivel ends, is zinc plated for corrosion resistance, and has a temperature rating of 250 degrees Fahrenheit. The overall length is adjustable from 3 inches to 6 inches by rotating the body. This turnbuckle is designed for use in structural applications to tension wire rope, rod and pipe.
Catalog No. 50-276-7227 Supplier Grainger Supplier No. 16X752
The Loos & Co. Turnbuckle Body is a forged steel turnbuckle with a body diameter of 1-1/16 inches. It has a working load limit of 3,200 pounds and meets ASTM A668 specifications. The turnbuckle has a drop forged body with swivel ends, is zinc plated for corrosion resistance, and has a temperature rating of 250 degrees Fahrenheit. The overall length is adjustable from 3 inches to 6 inches by rotating the body. This turnbuckle is designed for use in structural applications to tension wire rope, rod and pipe.