Learn More
Grainger Turnbuckle Body 1600 lb Load Cap 2 1/4 L
Catalog No. 502767088
Encompass_Preferred
The Grainger website features a variety of turnbuckles used for tensioning wire rope and cable. They offer several types including eye turnbuckles, hook turnbuckles, jaw turnbuckles, and take-up turnbuckles. The turnbuckles are available in various load capacities and materials including steel, stainless steel, and galvanized steel. Key specifications provided include thread size, overall length, and working load limit. The turnbuckles can be used in applications like securing loads, bracing structures, and pre-tensioning guy wires. Grainger provides technical support to help customers select the right turnbuckle for their application.
Catalog No. 50-276-7088 Supplier Grainger Supplier No. 16X751
The Grainger website features a variety of turnbuckles used for tensioning wire rope and cable. They offer several types including eye turnbuckles, hook turnbuckles, jaw turnbuckles, and take-up turnbuckles. The turnbuckles are available in various load capacities and materials including steel, stainless steel, and galvanized steel. Key specifications provided include thread size, overall length, and working load limit. The turnbuckles can be used in applications like securing loads, bracing structures, and pre-tensioning guy wires. Grainger provides technical support to help customers select the right turnbuckle for their application.