Learn More
Grainger Turnbuckle Body 4 600 lb Load Cap 4 in L
Catalog No. 502768037
Encompass_Preferred
The Grainger website features a variety of swivels and turnbuckles for lifting and rigging applications. These hardware accessories allow for rotation and adjustment of cables, chains, and slings. Offerings include single and double swivel hoist rings, swivel hooks, swivel shackles, clevis swivel shackles, turnbuckles, eye bolts, and more. Materials include carbon steel, stainless steel, galvanized steel, and alloy steel. Load capacities range from 1/4 ton to over 100 tons. Swivels provide 360 degree rotation while turnbuckles allow for adjustable length. These products meet ASME standards and are designed for overhead lifting, rigging, and load securement.
Catalog No. 50-276-8037 Supplier Grainger Supplier No. 16X756
The Grainger website features a variety of swivels and turnbuckles for lifting and rigging applications. These hardware accessories allow for rotation and adjustment of cables, chains, and slings. Offerings include single and double swivel hoist rings, swivel hooks, swivel shackles, clevis swivel shackles, turnbuckles, eye bolts, and more. Materials include carbon steel, stainless steel, galvanized steel, and alloy steel. Load capacities range from 1/4 ton to over 100 tons. Swivels provide 360 degree rotation while turnbuckles allow for adjustable length. These products meet ASME standards and are designed for overhead lifting, rigging, and load securement.